Få mere at vide
PF-04217903, MedChemExpress
Artikelnummer. 16495696
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
50 mg
100 mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16495696 100 mg
16477667 5 mg
16474918 10 mg
16471167 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16495696 Leverandør Medchem Express Leverandørnr. HY12017
MedChemExpress PF-04217903 is a potent ATP-competitive c-Met kinase inhibitor with Ki of 4.8 nM for human c-Met. PF-04217903 shows more than 1,000-fold selectivity relative to 208 kinases. Antiangiogenic properties.
PF-04217903 is a potent ATP-competitive c-Met kinase inhibitor with Ki of 4.8 nM for human c-Met. PF-04217903 shows more than 1,000-fold selectivity relative to 208 kinases. Antiangiogenic properties.
Tekniske data
Kemisk navn eller materiale PF-04217903
Fysisk form Solid
CAS 956905-27-4
Farve Off-White
Mængde 100 mg
Renhedsgrad noter Research
Molekylær formel C19H16N8O
Til brug med (applikation) Cancer-Kinase/protease
Opløselighedsinformation DMSO : 20 mg/mL (53.71 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (insoluble)
SMIL OCCN1N=CC(C2=NC3=C(N=C2)N=NN3CC4=CC5=CC=CN=C5C=C4)=C1
Molekylvægt (g/mol) 372.38
Formel vægt 372.38
Procent renhed 99.4%