missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Pexacerfont, MedChemExpress
Description
Pexacerfont is a selective corticotropin-releasing factor (CRF1) receptor antagonist with IC50 of 6.1±0.6 nM for human CRF1 receptor.
Specifications
Specifications
| Kemisk navn eller materiale | Pexacerfont |
| CAS | 459856-18-9 |
| Farve | White |
| Renhedsgrad noter | Research |
| Molekylær formel | C18H24N6O |
| Mængde | 5 mg |
| Synonym | BMS-562086 |
| Opløselighedsinformation | DMSO : 50 mg/mL (146.88 mM; Need ultrasonic) |
| SMIL | CC[C@H](NC1=NC(C)=NC2=C(C3=CC=C(OC)N=C3C)C(C)=NN21)C |
| Molekylvægt (g/mol) | 340.42 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?