Få mere at vide
Scharlab PETR.ETER 1L KP 40-60 PA
Brand:  Scharlab ET00921000
PETR.ETER 1L KP 40-60 PA
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Produktkode 11358644
  • ONLINE EXCLUSIVE PRICE
    668.85 DKK
    386.69 DKK / 1 L
    Save 282.16 DKK
    42% Slukket