Få mere at vide
Pesticide Mixture B in Dichloromethane, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Azinphos-methyl∣Bromacil∣Carboxin∣Chlorpyrifos∣Dimethoate∣Disulfoton∣Diuron∣Ethalfluralin∣Ethion∣Fenoxaprop-P-ethyl (R-enantiomer)∣Linuron∣Malathion∣Methomyl∣Napropamide∣Phorate∣Pyridaben∣Terbufos∣Thiamethoxam∣Vinclozolin |
| CAS | 86-50-0, 314-40-9, 5234-68-4, 2921-88-2, 60-51-5, 298-04-4 |
| Analyt- eller komponentnavne | Azinphos-methyl∣Bromacil∣Carboxin∣Chlorpyrifos∣Dimethoate∣Disulfoton∣Diuron∣Ethalfluralin∣Ethion∣Fenoxaprop-P-ethyl (R-enantiomer)∣Linuron∣Malathion∣Methomyl∣Napropamide∣Phorate∣Pyridaben∣Terbufos∣Thiamethoxam∣Vinclozolin |
| Forsendelsestilstand | +20°C |
| Certificeringer/compliance | ISO 17034 |
| Opløsningsmiddel eller Matrix | Dichloromethane |
| Mængde | 4 x 1 ml |
| Koncentration eller sammensætning (efter analyt eller komponenter) | Chlorpyrifos 50 μg/mL∣Dimethoate 50 μg/mL∣Ethalfluralin 50 μg/mL∣Phorate 50 μg/mL∣Vinclozolin 50 μg/mL∣Bromacil 250 μg/mL∣Fenoxaprop-P-ethyl (R-enantiomer) 250 μg/mL∣Linuron 250 μg/mL∣Malathion 250 μg/mL∣Napropamide 250 μg/mL∣Pyridaben 250 μg/mL∣Terbufos 250 μg/mL∣Thiamethoxam 250 μg/mL∣Azinphos-methyl 1000 μg/mL∣Carboxin 1000 μg/mL∣Disulfoton 1000 μg/mL∣Diuron 1000 μg/mL∣Ethion 1000 μg/mL∣Methomyl 1000 μg/mL |