missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Pesticide Mixture A in Dichloromethane, Dr. Ehrenstorfer
Specifications
Specifications
| Kemisk navn eller materiale | Aldicarb∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣Atrazine-desethyl∣Atrazine-desisopropyl∣Chlorothalonil∣Cyanazine∣Diazinon∣Dieldrin∣Ethofumesate∣gamma-HCH∣Metalaxyl∣Methoxychlor∣Metolachlor∣Oxycarboxin∣Parathion-ethyl∣Simazine∣Triallate |
| CAS | 116-06-3, 309-00-2, 959-98-8, 319-84-6, 6190-65-4, 1007-28-9 |
| Analyt- eller komponentnavne | Aldicarb∣Aldrin∣alpha-Endosulfan∣alpha-HCH∣Atrazine-desethyl∣Atrazine-desisopropyl∣Chlorothalonil∣Cyanazine∣Diazinon∣Dieldrin∣Ethofumesate∣gamma-HCH∣Metalaxyl∣Methoxychlor∣Metolachlor∣Oxycarboxin∣Parathion-ethyl∣Simazine∣Triallate |
| Forsendelsestilstand | +20°C |
| Certificeringer/compliance | ISO 17034 |
| Opløsningsmiddel eller Matrix | Dichloromethane |
| Mængde | 4 x 1 ml |
| Koncentration eller sammensætning (efter analyt eller komponenter) | Aldrin 200 μg/mL∣alpha-Endosulfan 200 μg/mL∣alpha-HCH 200 μg/mL∣Chlorothalonil 200 μg/mL∣Diazinon 200 μg/mL∣Dieldrin 200 μg/mL∣Ethofumesate 200 μg/mL∣gamma-HCH 200 μg/mL∣Metalaxyl 200 μg/mL∣Metolachlor 200 μg/mL∣Parathion-ethyl 200 μg/mL∣Simazine 200 μg/mL∣Triallate 200 μg/mL∣Atrazine-desethyl 1000 μg/mL∣Atrazine-desisopropyl 1000 μg/mL∣Cyanazine 1000 μg/mL∣Methoxychlor 1000 μg/mL∣Oxycarboxin 1000 μg/mL∣Aldicarb 4000 μg/mL |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?