Få mere at vide
Perzinfotel, MedChemExpress
Artikelnummer. 16420168
Encompass_Preferred
Skift visning
Mængde:
5 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
16420168 5 mg
16407708 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 16420168 Leverandør Medchem Express Leverandørnr. HY19168
MedChemExpress Perzinfotel (EAA-090) is a potent, selective, and competitive NMDA receptor antagonist with neuroprotective effects. Perzinfotel (EAA-090) shows high affinity (IC50=30 nM) for the glutamate site.
Perzinfotel (EAA-090) is a potent, selective, and competitive NMDA receptor antagonist with neuroprotective effects. Perzinfotel (EAA-090) shows high affinity (IC50=30 nM) for the glutamate site.
Tekniske data
Kemisk navn eller materiale Perzinfotel
Fysisk form Powder
CAS 144912-63-0
Farve Off-White
Mængde 5 mg
Renhedsgrad noter Research
Molekylær formel C9H13N2O5P
Til brug med (applikation) Neuroscience-Neurodegeneration
Synonym EAA-090
Opløselighedsinformation DMSO : 20.83 mg/mL (80.06 mM; Need ultrasonic) ∣H2O : 5.88 mg/mL (22.60 mM; ultrasonic and warming and heat to 60°C)
SMIL O=C(C1=C2NCCCN1CCP(O)(O)=O)C2=O
Molekylvægt (g/mol) 260.18
Formel vægt 260.18
Procent renhed 98.0%