Få mere at vide
Perylene-d12, TRC
Artikelnummer. 17998449
Skift visning
Mængde:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17998449 25 mg
17919019 10 mg
17928319 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 17998449 Leverandør Toronto Research Chemicals Leverandørnr. TRCP29130225MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1520-96-3
Molekylær formel C20 2H12
Molekylvægt (g/mol) 264.38
Synonym Perylene D12, Perylene-1,2,3,4,5,6,7,8,9,10,11,12-d12, Perylene-d12 (7CI,8CI,9CI), Perdeuterioperylene, [2H12]Perylene
IUPAC navn 1,2,3,4,5,6,7,8,9,10,11,12-dodecadeuterioperylene
SMIL [2H]c1c([2H])c2c([2H])c([2H])c([2H])c3c4c([2H])c([2H])c([2H])c5c([2H])c([2H])c([2H])c(c(c1[2H])c23)c45
Tekniske data
Kemisk navn eller materiale Perylene D12
Mængde 25 mg
InChI formel InChI=1S/C20H12/c1-5-13-6-2-11-17-18-12-4-8-14-7-3-10-16(20(14)18)15(9-1)19(13)17/h1-12H/i1D,2D,3D,4D,5D,6D,7D,8D,9D,10D,11D,12D
Formel vægt 264.1692