Få mere at vide
Perseitol, TRC
Artikelnummer. 30412582
Skift visning
Mængde:
50 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30412582 50 mg
30411511 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30412582 Leverandør Toronto Research Chemicals Leverandørnr. TRCP69822850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perseitol
CAS 527-06-0
Molekylær formel C7H16O7
Mængde 50 mg
InChI formel InChI=1S/C7H16O7/c8-1-3(10)5(12)7(14)6(13)4(11)2-9/h3-14H,1-2H2/t3-,4+,5-,6-,7+/m1/s1
SMIL [C@H]([C@@H]([C@H](CO)O)O)([C@@H]([C@@H](CO)O)O)O
IUPAC navn (2S,3R,5R,6R)-heptane-1,2,3,4,5,6,7-heptol
Molekylvægt (g/mol) 212.19
Formel vægt 212.0896