Få mere at vide
Perphenylcyclopentasilane, TRC
Artikelnummer. 30424327
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30424327 500 mg
30422267 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30424327 Leverandør Toronto Research Chemicals Leverandørnr. TRCP291110500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perphenylcyclopentasilane
CAS 1770-54-3
Molekylær formel C60H50Si5
Mængde 500 mg
Synonym Decaphenylcyclopentasilane, NSC 526968, Ogsol SI 30-10, 1,1,2,2,3,3,4,4,5,5-Decaphenylcyclopentasilane
InChI formel InChI=1S/C60H50Si5/c1-11-31-51(32-12-1)61(52-33-13-2-14-34-52)62(53-35-15-3-16-36-53,54-37-17-4-18-38-54)64(57-43-23-7-24-44-57,58-45-25-8-26-46-58)65(59-47-27-9-28-48-59,60-49-29-10-30-50-60)63(61,55-39-19-5-20-40-55)56-41-21-6-22-42-56/h1-50H
SMIL c1ccc(cc1)[Si]2(c3ccccc3)[Si](c4ccccc4)(c5ccccc5)[Si](c6ccccc6)(c7ccccc7)[Si](c8ccccc8)(c9ccccc9)[Si]2(c%10ccccc%10)c%11ccccc%11
IUPAC navn 1,1,2,2,3,3,4,4,5,5-decakis-phenylpentasilolane
Molekylvægt (g/mol) 911.47
Formel vægt 910.2759