Få mere at vide
Perphenazine N1-Oxide, Mikromol™
Artikelnummer. 30049583
Skift visning
Mængde:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049583 25 mg
17943076 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30049583 Leverandør Mikromol Leverandørnr. MM0826.030025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Perphenazine N1-Oxide
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perphenazine N1-Oxide
CAS 803615-00-1
Analyt- eller komponentnavne Perphenazine N1-Oxide
Forsendelsestilstand Room Temperature
Molekylær formel C21 H26 Cl N3 O2 S
Certificeringer/compliance ISO 9001
Synonym 4-[3-(2-Chloro-10H-phenothiazin-10-yl)propyl]-1-piperazineethanol 1-Oxide, 1-Piperazineethanol, 4-[3-(2-chlorophenothiazin-10-yl)propyl]-, 1-oxide (8CI), Perphenazine N1-Oxide
InChI formel InChI=1S/C21H26ClN3O2S/c22-17-6-7-21-19(16-17)24(18-4-1-2-5-20(18)28-21)9-3-8-23-10-12-25(27,13-11-23)14-15-26/h1-2,4-7,16,26H,3,8-15H2
SMIL OCC[N+]1([O-])CCN(CCCN2c3ccccc3Sc4ccc(Cl)cc24)CC1
IUPAC navn 2-[4-[3-(2-chlorophenothiazin-10-yl)propyl]-1-oxidopiperazin-1-ium-1-yl]ethanol
Molekylvægt (g/mol) 419.97
Mængde 25 mg
Formel vægt 419.1434 g/mol