Få mere at vide
Perindopril Diketopiperazine, TRC
Artikelnummer. 30003777
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30003777 50 mg
30012069 10 mg
30011853 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30003777 Leverandør Toronto Research Chemicals Leverandørnr. TRCP28752550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 129970-98-5
Molekylær formel C19 H30 N2 O4
Molekylvægt (g/mol) 350.45
Synonym (αS,3S,5aS,9aS,10aS)-decahydro-3-methyl-1,4-dioxo-α-propyl-Pyrazino[1,2-a]indole-2(1H)-acetic acid ethyl ester, Pyrazino[1,2-a]indole-2(1H)-acetic acid, decahydro-3-methyl-1,4-dioxo-α-propyl-, ethyl ester, [3S-[2(R*),3α,5aβ,9aβ,10aβ]]- (9CI), Perindopril Related Compound F (USP)
IUPAC navn ethyl (2S)-2-[(3S,5aS,9aS,10aS)-3-methyl-1,4-dioxo-5a,6,7,8,9,9a,10,10a-octahydro-3H-pyrazino[1,2-a]indol-2-yl]pentanoate
SMIL CCC[C@H](N1[C@@H](C)C(=O)N2[C@H]3CCCC[C@H]3C[C@H]2C1=O)C(=O)OCC
Tekniske data
Kemisk navn eller materiale (S)-ethyl 2-{(3S,5aS,9aS,10aS)-3-methyl-1,4-dioxodecahydropyrazino[1,2-a]indol-2(1H)-yl}pentanoate
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C19H30N2O4/c1-4-8-15(19(24)25-5-2)20-12(3)17(22)21-14-10-7-6-9-13(14)11-16(21)18(20)23/h12-16H,4-11H2,1-3H3/t12-,13-,14-,15-,16-/m0/s1
Formel vægt 350.2206
Procent renhed >95%