Få mere at vide
Perindopril-D4, TRC
Artikelnummer. 30421813
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30421813 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30421813 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2875041MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perindopril-D4
CAS 1356929-58-2
Renhedsgrad noter HPLC
Molekylær formel C19H28D4N2O5
Mængde 1 mg
Synonym [2S-[1[R*(R*)],2α,3aβ,7aβ]]-1-[2-[[1-(Ethoxycarbonyl)butyl]amino]-1-oxo-2,3,3,3-tetradeuteriopropyl]octahydro-1H-indole-2-carboxylic acid, (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl-2,3,3,3-d4]octahydro-1H-indole-2-carboxylic acid, Perindopril-d4, Perindopril-2',3',3',3'-D4
InChI formel InChI=1S/C19H32N2O5/c1-4-8-14(19(25)26-5-2)20-12(3)17(22)21-15-10-7-6-9-13(15)11-16(21)18(23)24/h12-16,20H,4-11H2,1-3H3,(H,23,24)/t12-,13-,14-,15-,16-/m0/s1/i3D3,12D
SMIL [2H]C([2H])([2H])[C@]([2H])(N[C@@H](CCC)C(=O)OCC)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)O
Molekylvægt (g/mol) 372.49
Formel vægt 372.2562
Procent renhed >95