Få mere at vide
Perindopril-d4 t-Butylamine Salt, TRC
Artikelnummer. 30412252
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30412252 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30412252 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2875071MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perindopril-d4 t-Butylamine Salt
CAS 1356929-59-3
Molekylær formel C19 2H4 H28 N2 O5 . C4 H11 N
Mængde 1 mg
Synonym [2S-[1[R*(R*)],2α,3aβ,7aβ]]-1-[2-[[1-(Ethoxycarbonyl)butyl]amino]-1-oxo-2,3,3,3-tetradeuteriopropyl]octahydro-1H-indole-2-carboxylic acid compd. with 2-methyl-2-propanamine (1:1), 2-Methylpropan-2-amine (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(ethoxycarbonyl)butyl]amino]-1-oxopropyl-2,3,3,3-d4]octahydro-1H-indole-2-carboxylate, Perindopril-d4 tert-butylamine, Perindopril-2',3',3',3'-D4 tert-butylamine, Perindopril-d4 erbumine, Perindopril-2',3',3',3'-D4 erbumine
InChI formel InChI=1S/C19H32N2O5.C4H11N/c1-4-8-14(19(25)26-5-2)20-12(3)17(22)21-15-10-7-6-9-13(15)11-16(21)18(23)24;1-4(2,3)5/h12-16,20H,4-11H2,1-3H3,(H,23,24);5H2,1-3H3/t12-,13-,14-,15-,16-;/m0./s1/i3D3,12D;
SMIL C(C)(C)(C)N.C([C@@](N[C@H](C(OCC)=O)CCC)(C([2H])([2H])[2H])[2H])(=O)N1[C@@]2([C@](C[C@H]1C(O)=O)(CCCC2)[H])[H]
IUPAC navn (2S,3aS,7aS)-1-[(2S)-2,3,3,3-tetradeuterio-2-[[(2S)-1-ethoxy-1-oxopentan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylic acid;2-methylpropan-2-amine
Molekylvægt (g/mol) 445.63
Formel vægt 445.3454