missing translation for 'onlineSavingsMsg'
Learn More

Perindopril Benzyl Ester, TRC

Product Code. 30419055
Change view
Click to view available options
Mængde:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30419055 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30419055 Supplier Toronto Research Chemicals Supplier No. TRCP287515500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Perindopril Benzyl Ester
CAS 122454-52-8
Molekylær formel C26H38N2O5
Mængde 500 mg
Synonym (2S,3aS,7aS)-1-[(2S)-2-[[(1S)-1-(Ethoxycarbonyl)butyl]amino]-1-oxopropyl]octahydro-1H-indole-2-carboxylic Acid Benzyl Ester
InChI formel InChI=1S/C26H38N2O5/c1-4-11-21(25(30)32-5-2)27-18(3)24(29)28-22-15-10-9-14-20(22)16-23(28)26(31)33-17-19-12-7-6-8-13-19/h6-8,12-13,18,20-23,27H,4-5,9-11,14-17H2,1-3H3/t18-,20-,21-,22-,23-/m0/s1
SMIL CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)OCc3ccccc3)C(=O)OCC
IUPAC navn benzyl (2S,3aS,7aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxopentan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carboxylate
Molekylvægt (g/mol) 458.59
Formel vægt 458.2781
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.