Få mere at vide
Perindopril Acyl-Beta-D-glucuronide, TRC
Artikelnummer. 30413302
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30413302 1 mg
30422422 10 mg
30420374 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30413302 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2875101MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 120398-66-5
Molekylær formel C25 H40 N2 O11
Molekylvægt (g/mol) 544.59
Synonym Perindopril Acyl-B-D-Glucuronide
IUPAC navn (2S,3S,4S,5R,6S)-6-[(2S,3aS,7aS)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxopentan-2-yl]amino]propanoyl]-2,3,3a,4,5,6,7,7a-octahydroindole-2-carbonyl]oxy-3,4,5-trihydroxyoxane-2-carboxylic acid
SMIL CCC[C@H](N[C@@H](C)C(=O)N1[C@H]2CCCC[C@H]2C[C@H]1C(=O)O[C@@H]3O[C@@H]([C@@H](O)[C@H](O)[C@H]3O)C(=O)O)C(=O)OCC
Tekniske data
Kemisk navn eller materiale Perindopril Acyl-beta-D-glucuronide
Mængde 1 mg
InChI formel InChI=1S/C25H40N2O11/c1-4-8-14(23(34)36-5-2)26-12(3)21(31)27-15-10-7-6-9-13(15)11-16(27)24(35)38-25-19(30)17(28)18(29)20(37-25)22(32)33/h12-20,25-26,28-30H,4-11H2,1-3H3,(H,32,33)/t12-,13-,14-,15-,16-,17-,18-,19+,20-,25-/m0/s1
Formel vægt 544.2632