Få mere at vide
Perilla Ketone, TRC
Artikelnummer. 30418096
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418096 25 mg
30417748 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30418096 Leverandør Toronto Research Chemicals Leverandørnr. TRCP28705025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 553-84-4
Molekylær formel C10H14O2
Molekylvægt (g/mol) 166.22
Synonym 1-(3-Furanyl)-4-methyl-1-pentanone, 1-(3-Furyl)-4-methyl-1-pentanone, 3-(4-Methylpentanoyl)furan, NSC 348407
IUPAC navn 1-(furan-3-yl)-4-methylpentan-1-one
SMIL CC(C)CCC(=O)c1cocc1
Tekniske data
Kemisk navn eller materiale Perilla Ketone
Mængde 25 mg
InChI formel InChI=1S/C10H14O2/c1-8(2)3-4-10(11)9-5-6-12-7-9/h5-8H,3-4H2,1-2H3
Formel vægt 166.0994