missing translation for 'onlineSavingsMsg'
Få mere at vide

Perilla Ketone, TRC

Artikelnummer. 30417748
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30417748 250 mg
30418096 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30417748 Leverandør Toronto Research Chemicals Leverandørnr. TRCP287050250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 553-84-4
Molekylær formel C10H14O2
Molekylvægt (g/mol) 166.22
Synonym 1-(3-Furanyl)-4-methyl-1-pentanone, 1-(3-Furyl)-4-methyl-1-pentanone, 3-(4-Methylpentanoyl)furan, NSC 348407
IUPAC navn 1-(furan-3-yl)-4-methylpentan-1-one
SMIL CC(C)CCC(=O)c1cocc1

Specifications

Kemisk navn eller materiale Perilla Ketone
Mængde 250 mg
InChI formel InChI=1S/C10H14O2/c1-8(2)3-4-10(11)9-5-6-12-7-9/h5-8H,3-4H2,1-2H3
Formel vægt 166.0994
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.