Få mere at vide
Tocris Bioscience™ Perifosine
GREENER_CHOICE
Artikelnummer. 18775999
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
18775999 50 mg
18723168 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 18775999 Leverandør Tocris Bioscience™ Leverandørnr. 6087/50
PKB/Akt inhibitor; also modulates Kv2.1 current
Perifosine is a PKB/Akt inhibitor; accumulates in and reorganizes lipid rafts disrupting Akt signaling. Also inhibits MAPK pathway and induces autophagy. Induces apoptosis in leukemia cell lines. Decreases Kv2.1 current amplitude by modulating its inactivation gating. Antitumor. Orally bioavailable.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perifosine
CAS 157716-52-4
Synonym Octadecyl-(1,1-dimethyl-4-piperidylio) phosphate
Mængde 50 mg
Opløselighed Soluble to 100mM in water and to 100mM in ethanol
Molekylær formel C25H52NO4P
Formel vægt 461.66
Fysisk form Solid
Farve White
Produkttype Bioactive Small Molecule
Renhed >98%
Vis mere Vis mindre
For Research Use Only.