Få mere at vide
Tocris Bioscience™ Perifosine
GREENER_CHOICE
PKB/Akt inhibitor; also modulates Kv2.1 current
Brand:  Tocris Bioscience™ 6087/50
Encompass
Produktkode 15717216
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Perifosine is a PKB/Akt inhibitor; accumulates in and reorganizes lipid rafts disrupting Akt signaling. Also inhibits MAPK pathway and induces autophagy. Induces apoptosis in leukemia cell lines. Decreases Kv2.1 current amplitude by modulating its inactivation gating. Antitumor. Orally bioavailable.
Tekniske data
Tekniske data
Perifosine
Octadecyl-(1,1-dimethyl-4-piperidylio) phosphate
Soluble to 100mM in water and to 100mM in ethanol
White
157716-52-4
50 mg
Solid
Bioactive Small Molecule
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner
Produkttitel
Vælg et problem
For Research Use Only.