missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perforin Antibody (PRF1/7077R), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-14154AF405
Additional Details : Weight : 0.00970kg
Description
Perforin Monoclonal antibody specifically detects Perforin in Human samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Perforin | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Antibody | |
IgG κ |
Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
PRF1/7077R | |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
Cytolysin, FLH2, HPLH2, HPLH2lymphocyte pore forming protein, Lymphocyte pore-forming protein, MGC65093, P1PFN1, perforin 1 (pore forming protein), perforin-1, PFPcytolysin | |
Recombinant human Perforin protein fragment (around aa 413-552) (exact sequence is proprietary) (Uniprot: P14222) | |
0.1 mL | |
Adaptive Immunity, Apoptosis, Immunology, Innate Immunity | |
5551 | |
Store at 4°C in the dark. | |
Purified |