missing translation for 'onlineSavingsMsg'
Få mere at vide

Perfluorotetradecanoic Acid, TRC

Artikelnummer. 30010211
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30010211 500 mg
30007906 5 g
30006874 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Artikelnummer. 30010211 Leverandør Toronto Research Chemicals Leverandørnr. TRCP286600500MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 376-06-7
Molekylær formel C14 H F27 O2
Molekylvægt (g/mol) 714.11
Synonym Tetradecanoic acid, heptacosafluoro- (9CI), 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-Heptacosafluorotetradecanoic acid, PFTeDA, Perfluoro-n-tetradecanoic acid, Perfluoromyristic acid, Perfluorotetradecanecarboxylic acid, Perfluorotetradecanoic acid
IUPAC navn 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,13,13,14,14,14-heptacosafluorotetradecanoic acid
SMIL OC(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F

Tekniske data

Kemisk navn eller materiale Perfluorotetradecanoic acid
Renhedsgrad noter HPLC
Mængde 500 mg
InChI formel InChI=1S/C14HF27O2/c15-2(16,1(42)43)3(17,18)4(19,20)5(21,22)6(23,24)7(25,26)8(27,28)9(29,30)10(31,32)11(33,34)12(35,36)13(37,38)14(39,40)41/h(H,42,43)
Formel vægt 713.9545
Procent renhed >95%
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.