missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluorooctylphosphoric Acid (>85%), TRC
Specifications
Specifications
| Kemisk navn eller materiale | Perfluorooctylphosphoric Acid |
| CAS | 40143-78-0 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C8 H2 F17 O3 P |
| Mængde | 25 mg |
| Synonym | Perfluorooctylphosphonic Acid |
| InChI formel | InChI=1S/C8H2F17O3P/c9-1(10,3(13,14)5(17,18)7(21,22)23)2(11,12)4(15,16)6(19,20)8(24,25)29(26,27)28/h(H2,26,27,28) |
| SMIL | OP(=O)(O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC navn | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,8-heptadecafluorooctylphosphonic acid |
| Molekylvægt (g/mol) | 500.0466 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?