Få mere at vide
Perfluorohexylphosphonic Acid, TRC
Artikelnummer. 30133132
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30133132 100 mg
30132980 50 mg
17964955 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133132 Leverandør Toronto Research Chemicals Leverandørnr. TRCP270615100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 40143-76-8
Molekylær formel C6 H2 F13 O3 P
Molekylvægt (g/mol) 400.03
Synonym Phosphonic acid, P-(1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexyl)-, Phosphonic acid, (tridecafluorohexyl)- (9CI), P-(1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluorohexyl)phosphonic acid, (Perfluorohexane)phosphonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluorohexyl phosphonic acid, Perfluorohexylphosphonic acid, Tridecafluorohexylphosphonic acid
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexylphosphonic acid
SMIL OP(=O)(O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale Perfluorohexylphosphonic acid
Renhedsgrad noter HPLC
Mængde 100 mg
InChI formel InChI=1S/C6H2F13O3P/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22/h(H2,20,21,22)
Formel vægt 399.9534
Procent renhed >95%