Få mere at vide
Perfluorohexanesulfonic Acid (Technical Grade), TRC
Artikelnummer. 30011719
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30011719 50 mg
30005919 100 mg
17981495 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30011719 Leverandør Toronto Research Chemicals Leverandørnr. TRCP99973850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 355-46-4
Molekylær formel C6 H F13 O3 S
Molekylvægt (g/mol) 400.11
Synonym 1-Hexanesulfonic acid, tridecafluoro- (6CI,8CI), 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-hexanesulfonic acid, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluorohexane-1-sulfonic acid, 1-Perfluorohexanesulfonic acid, PFHxS
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonic acid
SMIL OS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale Perfluorohexanesulfonic acid
Mængde 50 mg
InChI formel InChI=1S/C6HF13O3S/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22/h(H,20,21,22)
Formel vægt 399.9439