Få mere at vide
Perfluorohexane Sulphonyl Fluoride (>80%), TRC
Artikelnummer. 30133842
Skift visning
Mængde:
1 g
2.5 g
5 g
Pakningsstørrelse:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30133842 2.5 g 2.5g
30136124 5 g -
30008647 1 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30133842 Leverandør Toronto Research Chemicals Leverandørnr. TRCP9992502.5G
Denne vare kræver en prisopdatering, inden den kan bestilles.
Anmod om en pris på denne vare her.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 423-50-7
Molekylær formel C6F14O2S
Molekylvægt (g/mol) 402.11
Synonym 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluoro-1-hexanesulfonyl Fluoride, 1,1,2,2,3,3,4,4,5,5,6,6,6-Tridecafluorohexane-1-sulfonyl Fluoride, Perfluoro-1-hexanesulfonyl Fluoride, Perfluorohexanesulfonyl Fluoride, Perfluorohexylsulfonyl Fluoride, Tridecafluorohexanesulfonyl Fluoride
IUPAC navn 1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane-1-sulfonyl fluoride
SMIL FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)S(=O)(=O)F
Tekniske data
Kemisk navn eller materiale Perfluorohexane Sulphonyl Fluoride
Mængde 2.5 g
InChI formel InChI=1S/C6F14O2S/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)23(20,21)22
Formel vægt 401.9395