Få mere at vide
Perfluorobutylsulphonamide, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Perfluorobutane Sulfonamide |
| CAS | 30334-69-1 |
| Renhedsgrad noter | HPLC |
| Molekylær formel | C4 H2 F9 N O2 S |
| Mængde | 500 mg |
| Synonym | Perfluorobutane Sulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-n-butanesulfonamide, Nonafluorobutanesulfonamide, Perfluorobutanesulfonamide, n-Nonafluorobutanesulfonamide, FBSA, FBSA, Perfluorobutane Sulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-1-butanesulfonamide, 1,1,2,2,3,3,4,4,4-Nonafluoro-n-butanesulfonamide, Nonafluorobutanesulfonamide, Perfluorobutanesulfonamide, n-Nonafluorobutanesulfonamide, PFBSA |
| InChI formel | InChI=1S/C4H2F9NO2S/c5-1(6,3(9,10)11)2(7,8)4(12,13)17(14,15)16/h(H2,14,15,16) |
| SMIL | NS(=O)(=O)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F |
| IUPAC navn | 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonamide |
| Molekylvægt (g/mol) | 299.11 |
| Vis mere |