Få mere at vide
Perfluoro-octylethanoic Acid, TRC
Artikelnummer. 17984955
Skift visning
Mængde:
50 mg
100 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17984955 50 mg
17973795 250 mg
17943645 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 17984955 Leverandør Toronto Research Chemicals Leverandørnr. TRCP28625850MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 27854-31-5
Molekylær formel C10 H3 F17 O2
Molekylvægt (g/mol) 478.1
Synonym Decanoic acid, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluoro-, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Heptadecafluorodecanoic acid, 8:2 FTCA, 8:2 Fluorotelomer carboxylic acid, Perfluoro-octylethanoic acid, 2H,2H-Perfluorodecanoic acid
IUPAC navn 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodecanoic acid
SMIL OC(=O)CC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
Tekniske data
Kemisk navn eller materiale 2H,2H-Perfluorodecanoic Acid
Mængde 50 mg
InChI formel InChI=1S/C10H3F17O2/c11-3(12,1-2(28)29)4(13,14)5(15,16)6(17,18)7(19,20)8(21,22)9(23,24)10(25,26)27/h1H2,(H,28,29)
Formel vægt 477.9862