missing translation for 'onlineSavingsMsg'
Få mere at vide

Perfluoro-n-pentane, TRC

Artikelnummer. 30418623
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30418623 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30418623 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2297151G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Dodecafluoropentane
CAS 678-26-2
Renhedsgrad noter GC
Molekylær formel C5 F12
Mængde 1 g
Synonym Perfluoropentane, Dodecafluoropentane, Pentane','1,1,1,2,2,3,3,4,4,5,5,5-dodecafluoro-, Pentane','dodecafluoro- (6CI,7CI,8CI,9CI), 1,1,1,2,2,3,3,4,4,5,5,5-Dodecafluoropentane, Dodecafluoropentane, FC 4112, FC 87, Fluorinert FC 87, Fluorinert PF 5050, Flutec PP 50, NVX 108, PF 5050, PFC 41-12, Perflenapent, Perfluoro-n-pentane, Perfluoropentane, QW 7437, R 41(12), R-41121,1,1,2,2,3,3,4,4,5,5,5-Dodecafluoropentane, Dodecafluoropentane, FC 4112, FC 87, Fluorinert FC 87, Fluorinert PF 5050, Flutec PP 50, NVX 108, PF 5050, PFC 41-12, Perflenapent, Perfluoro-n-pentane, Perfluoropentane, QW 7437, R 41(12), R-4112
InChI formel InChI=1S/C5F12/c6-1(7,2(8,9)4(12,13)14)3(10,11)5(15,16)17
SMIL FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)F
IUPAC navn 1,1,1,2,2,3,3,4,4,5,5,5-dodecafluoropentane
Molekylvægt (g/mol) 288.034
Formel vægt 287.981
Procent renhed >95
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.