missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoic Acid Methyl Ester, TRC
Specifications
Specifications
| Kemisk navn eller materiale | Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoic Acid Methyl Ester |
| CAS | 39187-47-8 |
| Molekylær formel | C13H3F23O5 |
| Mængde | 500 mg |
| Synonym | Propanoic acid','2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]-','methyl ester, Propanoic acid','2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(heptafluoropropoxy)propoxy]propoxy]-','methyl ester (9CI), Methyl perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoate |
| InChI formel | InChI=1S/C13H3F23O5/c1-38-2(37)3(14,7(19,20)21)39-12(33,34)5(17,9(25,26)27)41-13(35,36)6(18,10(28,29)30)40-11(31,32)4(15,16)8(22,23)24/h1H3 |
| SMIL | COC(=O)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(OC(F)(F)C(F)(F)C(F)(F)F)C(F)(F)F)C(F)(F)F)C(F)(F)F |
| IUPAC navn | methyl 2,3,3,3-tetrafluoro-2-[1,1,2,3,3,3-hexafluoro-2-[1,1,2,3,3,3-hexafluoro-2-(1,1,2,2,3,3,3-heptafluoropropoxy)propoxy]propoxy]propanoate |
| Molekylvægt (g/mol) | 676.12 |
| Formel vægt | 675.9613 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?