Learn More
Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers, tech., Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its application in the characterization of non-aqueous phase liquid/tracer interaction in support of a vadose zone partitioning interwell tracer test.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | -68°C |
Density | 1.888 |
Boiling Point | 125°C to 128°C |
Refractive Index | 1.297 |
Quantity | 10 g |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 450.07 |
Grade | Technical |
Chemical Name or Material | Perfluoro-1,3,5-trimethylcyclohexane, mixture of isomers |