Learn More
Cytiva Perfluor Rubber O-Ring
Product Code. 11390092 Shop All Cytiva Products
Change view
Click to view available options
Size:
1 x 3 mm
Unit Size:
Pack of 10
Product Code. Size Unit Size
11390092 1 x 3 mm Pack of 10
1 options
Product Code. 11390092 Supplier Cytiva Supplier No. 19003601
  • ONLINE EXCLUSIVE PRICE
    945.00 DKK
    878.71 DKK / Pakke med 10
    Spare 66.29 DKK
    7% Slukket
På lager
Specifications
For Use With (Application) Ettan LC
Quantity 10/Pk.
Product Type O-Ring
Material Perfluor Rubber
Size 1 x 3 mm