Få mere at vide
Peretinoin, TRC
Artikelnummer. 30425481
Skift visning
Mængde:
5 mg
10 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30425481 5 mg
30411186 50 mg
30418095 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30425481 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2861205MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 81485-25-8
Molekylær formel C20H30O2
Molekylvægt (g/mol) 302.45
Synonym (2E,4E,6E,10E)-3,7,11,15-Tetramethyl-2,4,6,10,14-hexadecapentaenoic Acid, (all-E)-3,7,11,15-Tetramethyl-2,4,6,10,14-hexadecapentaenoic Acid, 4,5-Didehydrogeranyl Geranoic Acid, Acyclic Retinoid, E 5166, NIK 333, Polyprenoic Acid, All-trans-3,7,11,15-Tetramethyl-2,4,6,10,14-hexadecapentaenoic Acid
IUPAC navn (2E,4E,6E,10E)-3,7,11,15-tetramethylhexadeca-2,4,6,10,14-pentaenoic acid
SMIL CC(=CCC\C(=C\CC\C(=C\C=C\C(=C\C(=O)O)\C)\C)\C)C
Tekniske data
Kemisk navn eller materiale Peretinoin
Mængde 5 mg
InChI formel InChI=1S/C20H30O2/c1-16(2)9-6-10-17(3)11-7-12-18(4)13-8-14-19(5)15-20(21)22/h8-9,11,13-15H,6-7,10,12H2,1-5H3,(H,21,22)/b14-8+,17-11+,18-13+,19-15+
Formel vægt 302.2246