missing translation for 'onlineSavingsMsg'
Learn More

Perchloric Acid, ACS Reagent, Honeywell Fluka™

Brand:  Honeywell Fluka 244252-4X2.5L

Additional Details : CAS Number : 7601-90-3 Weight : 16.60000kg Transport : UN number : 1873 Chem class : 5.1 Pack group : I

Quantity 4 x 2.5L

 View more versions of this product

Product Code. 15642650

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +45 70 27 99 20 for assistance


Specifications

Chemical Identifiers

7601-90-3
100.454
VLTRZXGMWDSKGL-UHFFFAOYSA-N
CHEBI:29221
OCl(=O)(=O)=O
ClHO4
MFCD00011325
24247
perchloric acid

Specifications

Perchloric Acid
4 x 2.5L
ACS Reagent
HClO4
1873
VLTRZXGMWDSKGL-UHFFFAOYSA-N
OCl(=O)(=O)=O
100.454
CHEBI:29221
≤0.003%
113°C (235.4°F)
Undesignated
7601-90-3
ClHO4
MFCD00011325
70%
1.664g/mL at 25°C
perchloric acid
24247
100.46g/mol
6.8mmHg (25°C)
Videos
SDS