Få mere at vide
Perchlorate Standard: ClO4- @ 10000 μg/mL in H2O, VHG™
Artikelnummer. 17908264
Skift visning
Mængde:
500 ml
Artikelnummer. Mængde
17908264 500 ml
1 options
Artikelnummer. 17908264 Leverandør VHG Labs Leverandørnr. VHGI1PCLO4500
VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications
Perchlorate Standard: ClO4- @ 10000 μg/mL in H2O
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perchlorate
CAS 14797-73-0
Metoder Ion Chromatography/Wet Chemistry
Analyt- eller komponentnavne Perchlorate
Molekylær formel (Cl O4)-
Synonym ClO4−
IUPAC navn Perchlorate
Opløsningsmiddel eller Matrix Water
Molekylvægt (g/mol) 99.45
Mængde 500 ml
Formel vægt 99.45 g/mol