Få mere at vide
Perazine Sulphoxide, LoGiCal
Artikelnummer. 30460460
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30460460 10 mg
1 options
Artikelnummer. 30460460 Leverandør LoGiCal Leverandørnr. LGCFOR0157.01
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perazine Sulphoxide
CAS 20627-44-5
Analyt- eller komponentnavne Perazine Sulphoxide
Forsendelsestilstand Room Temperature
Molekylær formel C20 H25 N3 O S
Certificeringer/compliance ISO 9001
Synonym Perazine Sulphoxide
InChI formel InChI=1S/C20H25N3OS/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)25(24)20-10-5-3-8-18(20)23/h2-5,7-10H,6,11-16H2,1H3
Løsningstype Neat
SMIL CN1CCN(CCCN2c3ccccc3S(=O)c4ccccc24)CC1
IUPAC navn 10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine 5-oxide
Industry Type Clinical Testing
Molekylvægt (g/mol) 355.5
Mængde 10 mg
Formel vægt 355.1718 g/mol