Få mere at vide
Perazine Dimaleate, Mikromol™
Artikelnummer. 30052594
Skift visning
Mængde:
100 mg
Artikelnummer. Mængde
30052594 100 mg
1 options
Artikelnummer. 30052594 Leverandør Mikromol Leverandørnr. MM0815.00
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Perazine Dimaleate
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perazine Dimaleate
CAS 14516-56-4
Analyt- eller komponentnavne Perazine Dimaleate
Forsendelsestilstand Room Temperature
Molekylær formel C20 H25 N3 S . 2 C4 H4 O4
Certificeringer/compliance ISO 17025
Synonym Perazine Dimaleate, 10H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, (Z)-2-butenedioate (1:2), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, dimaleate (6CI), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, maleate (1:2) (8CI), Perazine dimaleate, Perazine maleate, Pernazinum, Prochlorperazine Maleate Impurity B as Dimaleate, Prochlorperazine Impurity B as Dimaleate
InChI formel InChI=1S/C20H25N3S.2C4H4O4/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)24-20-10-5-3-8-18(20)23;2*5-3(6)1-2-4(7)8/h2-5,7-10H,6,11-16H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
SMIL CN1CCN(CCCN2c3ccccc3Sc4ccccc24)CC1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O
IUPAC navn (Z)-but-2-enedioic acid;10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
Molekylvægt (g/mol) 571.64
Mængde 100 mg
Formel vægt 571.1988 g/mol