Få mere at vide
Perazine Dimaleate, LoGiCal
Artikelnummer. 30457816
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30457816 10 mg
1 options
Artikelnummer. 30457816 Leverandør LoGiCal Leverandørnr. LGCFOR0815.00
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Perazine Dimaleate
CAS 14516-56-4
Analyt- eller komponentnavne Perazine Dimaleate
Forsendelsestilstand Room Temperature
Molekylær formel C20 H25 N3 S . 2 C4 H4 O4
Certificeringer/compliance ISO 9001
Synonym Perazine Dimaleate, 10H-Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, (Z)-2-butenedioate (1:2), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, dimaleate (6CI), Phenothiazine, 10-[3-(4-methyl-1-piperazinyl)propyl]-, maleate (1:2) (8CI), Perazine dimaleate, Perazine maleate, Pernazinum, Prochlorperazine Maleate Impurity B as Dimaleate, Prochlorperazine Impurity B as Dimaleate
InChI formel InChI=1S/C20H25N3S.2C4H4O4/c1-21-13-15-22(16-14-21)11-6-12-23-17-7-2-4-9-19(17)24-20-10-5-3-8-18(20)23;2*5-3(6)1-2-4(7)8/h2-5,7-10H,6,11-16H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1-
Løsningstype Neat
SMIL CN1CCN(CCCN2c3ccccc3Sc4ccccc24)CC1.OC(=O)\C=C/C(=O)O.OC(=O)\C=C/C(=O)O
IUPAC navn (Z)-but-2-enedioic acid;10-[3-(4-methylpiperazin-1-yl)propyl]phenothiazine
Industry Type Clinical Testing
Molekylvægt (g/mol) 571.64
Mængde 10 mg
Formel vægt 571.1988 g/mol