missing translation for 'onlineSavingsMsg'
Få mere at vide

Pepstatin, MedChemExpress

Product Code. 16474957
Encompass_Preferred
Change view
Click to view available options
Mængde:
10 mg
50 mg
Unit Size:
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde unitSize
16474957 50 mg 50mg
16479318 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16474957 Supplier Medchem Express Supplier No. HYP0018

Venligst for at købe denne vare. Brug for en webkonto? Register dig hos os i dag!

This item is not returnable. View return policy

MedChemExpress Pepstatin (Pepstatin A) is a specific aspartic protease inhibitor produced by actinomycetes, with IC50s of 4.5 nM, 6.2 nM, 150 nM, 290 nM, 520 nM and 260 nM for hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease and hemoglobin-acid protease, respectively. Pepstatin Ammonium also inhibits HIV protease.

Pepstatin (Pepstatin A) is a specific aspartic protease inhibitor produced by actinomycetes, with IC50s of 4.5 nM, 6.2 nM, 150 nM, 290 nM, 520 nM and 260 nM for hemoglobin-pepsin, hemoglobin-proctase, casein-pepsin, casein-proctase, casein-acid protease and hemoglobin-acid protease, respectively. Pepstatin Ammonium also inhibits HIV protease.

Specifications

Kemisk navn eller materiale Pepstatin
Fysisk form Solid
CAS 26305-03-3
Farve White
Mængde 50 mg
Renhedsgrad noter Research
Molekylær formel C34H63N5O9
Til brug med (applikation) COVID-19-anti-virus
Synonym Pepstatin A
Opløselighedsinformation DMSO : 41.67 mg/mL (60.75 mM; Need ultrasonic)
SMIL CC(C[C@H](NC([C@@H](NC([C@@H](NC(CC(C)C)=O)C(C)C)=O)C(C)C)=O)[C@@H](O)CC(N[C@@H](C)C(N[C@@H](CC(C)C)[C@@H](O)CC(O)=O)=O)=O)C
Molekylvægt (g/mol) 685.89
Formel vægt 685.89
Procent renhed 98.0%
Grad Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.