missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen II Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1081.84 DKK - 3058.99 DKK
Specifications
Antigen | Pepsinogen II |
---|---|
Concentration | 0.22 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16816644
|
Proteintech
16881-1-AP-20UL |
20 ÎĽL |
1081.84 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16806644
|
Proteintech
16881-1-AP-150UL |
150 ÎĽL |
3058.99 DKK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Hydrolyzes a variety of proteins.Specifications
Pepsinogen II | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P04073, P20142, Q9D7R7 | |
109820, 24864, 5225 | |
Pepsinogen II Fusion Protein Ag10332 | |
Primary | |
-20°C | |
PGC |
0.22 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Gastricsin, Pepsinogen C, Pepsinogen II, PEPSINOGEN, GROUP II, PGC, Progastricsin, progastricsin (pepsinogen C) | |
PGC | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Pepsinogen II Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech