missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pepsinogen C/PGC/Progastricsin Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Bio-Techne NBP2-99924H
Additional Details : Weight : 0.00970kg
Description
Pepsinogen C/PGC/Progastricsin Polyclonal specifically detects Pepsinogen C/PGC/Progastricsin in Human samples. It is validated for ELISA.Specifications
Pepsinogen C/PGC/Progastricsin | |
Polyclonal | |
PBS | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA | |
HRP | |
EC 3.4.23, EC 3.4.23.3, FLJ99563, gastricsin, PEPC, pepsin C, Pepsinogen C, pepsinogen group II, PGII, preprogastricsin, progastricsin (pepsinogen C) | |
Produced in rabbits immunized with purified, recombinant Pepsinogen C/PGC/Progastricsin (NP_002621.1; Met 1-Ala 388). | |
0.1 mL | |
Phospho Specific | |
5225 | |
Store at 4°C in the dark. | |
IgG |