Få mere at vide
Pep 2-8, Tocris Bioscience™
GREENER_CHOICE
Potent PCSK9 inhibitor
Brand:  Tocris Bioscience 6423/1
Encompass
Produktkode 15953291
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter »
Denne vare kan ikke returneres. Se returpolitik
Beskrivelse
Beskrivelse
Pep 2-8 is a proprotein convertase subtilisin/kexin type 9 (PCSK9) inhibitor. Potent inhibitor of PCSK9 binding to LDL receptor (IC50 = 0.8 μM). Restores LDL uptake in HepG2 cells treated with PCSK9.
Tekniske data
Tekniske data
1541011-97-5
Store at -20°C
Observed MW: 1715.88
1 mg
Produktforslag
Produktforslag
Videoer
SDS
Dokumenter
Dokumenter
Certifikater
Kampagner
Kampagner