Få mere at vide
Pentylenetetrazole-d6, TRC
Artikelnummer. 30423918
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30423918 50 mg
30428673 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30423918 Leverandør Toronto Research Chemicals Leverandørnr. TRCP22750250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1329509-68-3
Molekylær formel C6H4D6N4
Molekylvægt (g/mol) 144.21
Synonym 6,7,8,9-Tetrahydro-5H-tetrazolo[1,5-a]azepine-d6, 1,2,3,3a-Tetrazacyclohepta-8a,2- cyclopentadiene-d6, 1,5-Pentamethylenetetrazole-d6, 6,7,8,9-Tetrahydro-5H- tetrazoloazepine-d6, 7,8,9,10-Tetrazabicyclo[5.3.0]-8,10-decadiene-d6, Angiazol-d6, Cardiazol-d6, Cardiazole-d6, Cardiol-d6, Korazole-d6, Leptazol-d6, Pentavet-d6, Pentazol-d6, Ventrazol-d6, Yetrazol-d6
SMIL [2H]C1([2H])C([2H])([2H])CC2=NN=NN2CC1([2H])[2H]
Tekniske data
Kemisk navn eller materiale Pentylenetetrazole-d6
Mængde 50 mg
InChI formel InChI=1S/C6H10N4/c1-2-4-6-7-8-9-10(6)5-3-1/h1-5H2/i1D2,2D2,3D2
Formel vægt 144.1282