missing translation for 'onlineSavingsMsg'
Få mere at vide

Pentyl-d11 Paraben, TRC

Product Code. 30414324
Change view
Click to view available options
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Mængde
30414324 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30414324 Supplier Toronto Research Chemicals Supplier No. TRCP2838025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale Pentyl-d11 Paraben
CAS 1216496-15-9
Molekylær formel C12H5D11O3
Mængde 5 mg
Synonym 4-Hydroxybenzoic Acid Pentyl-d11 Ester, 4-Hydroxybenzoic Acid n-Amyl-d11 Ester, Amyl-d11 p-Hydroxybenzoate, NSC 309817-d11, Pentyl-d11 4-Hydroxybenzoate, Pentyl-d11 p-Hydroxybenzoate
InChI formel InChI=1S/C12H16O3/c1-2-3-4-9-15-12(14)10-5-7-11(13)8-6-10/h5-8,13H,2-4,9H2,1H3/i1D3,2D2,3D2,4D2,9D2
SMIL [2H]C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])OC(=O)C1=CC=C(C=C1)O
IUPAC navn 1,1,2,2,3,3,4,4,5,5,5-undecadeuteriopentyl 4-hydroxybenzoate
Molekylvægt (g/mol) 219.32
Formel vægt 219.179
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.