missing translation for 'onlineSavingsMsg'
Learn More

Pentraxin 2/SAP Antibody (APCS/3240), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Bio-Techne NBP2-79848AF647

Product Code. 18245596

  • 4610.00 DKK / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pentraxin 2/SAP Monoclonal specifically detects Pentraxin 2/SAP in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pentraxin 2/SAP
Monoclonal
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
APCS
Recombinant fragment of human Pentraxin 2/SAP protein (around aa 143-223) (exact sequence is proprietary) (Uniprot: P02743)
0.1 mL
Neuroscience
Human
IgG2b κ
Immunohistochemistry, Immunohistochemistry (Paraffin)
APCS/3240
Immunohistochemistry, Immunohistochemistry-Paraffin
9.5S alpha-1-glycoprotein, amyloid P component, serum, MGC88159, PTX2serum amyloid P-component, SAPpentaxin-related, Serum Amyloid P component
Mouse
Protein A or G purified
RUO
Primary
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pentraxin 2/SAP Antibody (APCS/3240), Alexa Fluor™ 647, Novus Biologicals™ > 0.1 mL; Alexa Fluor 647

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.