Få mere at vide
Pentoxifylline-d6, TRC
Artikelnummer. 30414394
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30414394 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30414394 Leverandør Toronto Research Chemicals Leverandørnr. TRCP2765021MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentoxifylline-d6
CAS 1185878-98-1
Molekylær formel C13 2H6 H12 N4 O3
Mængde 1 mg
Synonym 1H-Purine-2,6-dione','3,7-dihydro-3,7-di(methyl-d3)-1-(5-oxohexyl)- (ACI), 3,7-Dihydro-3,7-di(methyl-d3)-1-(5-oxohexyl)-1H-purine-2,6-dione (ACI), Pentoxifylline-d6, Pentoxifylline-D6 (N,N-dimethyl-D6)
InChI formel InChI=1S/C13H18N4O3/c1-9(18)6-4-5-7-17-12(19)10-11(14-8-15(10)2)16(3)13(17)20/h8H,4-7H2,1-3H3/i2D3,3D3
SMIL [2H]C([2H])([2H])N1C(=O)N(CCCCC(=O)C)C(=O)c2c1ncn2C([2H])([2H])[2H]
IUPAC navn 1-(5-oxohexyl)-3,7-bis(trideuteriomethyl)purine-2,6-dione
Molekylvægt (g/mol) 284.34
Formel vægt 284.1756