Få mere at vide
Pentolinium Tartrate, LoGiCal
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Pentolinium Tartrate |
| CAS | 52-62-0 |
| Analyt- eller komponentnavne | Pentolinium Tartrate |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C15 H32 N2 . 2 C4 H5 O6 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Pyrrolidinium, 1,1'-(1,5-pentanediyl)bis[1-methyl-, (2R,3R)-2,3-dihydroxybutanedioate (1:2), 1,1'-Pentamethylenebis[1-methylpyrrolidinium hydrogen tartrate] (6CI), Pyrrolidinium, 1,1'-(1,5-pentanediyl)bis[1-methyl-, salt with (2R,3R)-2,3-dihydroxybutanedioic acid (1:2) (9CI), Pyrrolidinium, 1,1'-(1,5-pentanediyl)bis[1-methyl-, salt with [R-(R*,R*)]-2,3-dihydroxybutanedioic acid (1:2), Pyrrolidinium, 1,1'-pentamethylenebis[1-methyl-, tartrate (1:2) (8CI), Butanedioic acid, 2,3-dihydroxy- (2R,3R)-, ion(1-), 1,1'-(1,5-pentanediyl)bis[1-methylpyrrolidinium] (2:1) (9CI), Butanedioic acid, 2,3-dihydroxy-[R-(R*,R*)]-, ion(1-), 1,1'-(1,5-pentanediyl)bis[1-methylpyrrolidinium] (2:1), Tartaric acid, ion(1-), 1,1'-pentamethylenebis[1-methylpyrrolidinium] (2:1) (8CI), 1,1'-Pentamethylenebis(1-methylpyrrolidinium tartrate), Ansolysen, Ansolysen bitartrate, Ansolysen tartrate, M and B 2050A, Pendine, Pentapyrrolidinium bitartrate, Pentapyrrolidium bitartrate, Pentilium, Pentolinium bitartrate, Pentolinium tartrate, Pentolonium tartrate, Recuryl, Tensilest |
| InChI formel | InChI=1S/C15H32N2.2C4H6O6/c1-16(12-6-7-13-16)10-4-3-5-11-17(2)14-8-9-15-17;2*5-1(3(7)8)2(6)4(9)10/h3-15H2,1-2H3;2*1-2,5-6H,(H,7,8)(H,9,10)/q+2;;/p-2/t;2*1-,2-/m.11/s1 |
| Løsningstype | Neat |
| SMIL | C[N+]1(CCCCC[N+]2(C)CCCC2)CCCC1.O[C@H]([C@@H](O)C(=O)[O-])C(=O)O.O[C@H]([C@@H](O)C(=O)[O-])C(=O)O |
| Vis mere |