Få mere at vide
Pentanoic-4,4-d2 Acid, TRC
Artikelnummer. 30453017
Skift visning
Mængde:
2.5 mg
Artikelnummer. Mængde
30453017 2.5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453017 Leverandør Toronto Research Chemicals Leverandørnr. TRCV0914182.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentanoic-4,4-d2 Acid
CAS 83741-75-7
Molekylær formel C5 2H2 H8 O2
Mængde 2.5 mg
Synonym Pentanoic-4,4-d2 acid (9CI), γ-Dideuteropentanoic acid, Pentanoic acid-4,4-d2, Valeric acid-d2, Valeric-γ,γ-d2 acid, Pentanoic-4,4-D2 acid
InChI formel InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i2D2
SMIL [2H]C([2H])(C)CCC(=O)O
IUPAC navn 4,4-dideuteriopentanoic acid
Molekylvægt (g/mol) 104.144
Formel vægt 104.081