Få mere at vide
Pentanoic-2,2-d2 Acid, TRC
Artikelnummer. 30452672
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30452672 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30452672 Leverandør Toronto Research Chemicals Leverandørnr. TRCV09141610MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentanoic-2,2-d2 Acid
CAS 64118-37-2
Molekylær formel C5 2H2 H8 O2
Mængde 10 mg
Synonym Pentanoic-2,2-d2 acid (9CI), α-Dideuteropentanoic acid, Pentanoic acid-2,2-d2, Valeric acid-d2, Valeric-α,α-d2 acid, Pentanoic-2,2-D2 acid
InChI formel InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i4D2
SMIL [2H]C([2H])(CCC)C(=O)O
IUPAC navn 2,2-dideuteriopentanoic acid
Molekylvægt (g/mol) 104.144
Formel vægt 104.081