Få mere at vide
Pentanoic-2,2,3,3-d4 Acid, TRC
Artikelnummer. 30453475
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30453475 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30453475 Leverandør Toronto Research Chemicals Leverandørnr. TRCV91417150MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentanoic-2,2,3,3-d4 Acid
CAS 1219804-71-3
Molekylær formel C5 D4 H6 O2
Mængde 50 mg
Synonym Pentanoic-2,2,3,3-d4 acid, Pentanoic acid-2,2,3,3-d4, Valeric acid-d4, Valeric-α,α,β,β-d4 acid, Pentanoic-2,2,3,3-D4 acid
InChI formel InChI=1S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i3D2,4D2
SMIL [2H]C([2H])(CC)C([2H])([2H])C(=O)O
IUPAC navn 2,2,3,3-tetradeuteriopentanoic acid
Molekylvægt (g/mol) 106.156
Formel vægt 106.093