Få mere at vide
Pentanoic-2,2,3,3-d4 Acid, CDN
Artikelnummer. 30311127
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30311127 0.1 g 0.1g
30326689 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311127 Leverandør CDN Leverandørnr. CDND60850.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Pentanoic-2,2,3,3-d4 Acid
CAS 1219804-71-3
Molekylær formel C5 D4 H6 O2
Mængde 0.1 g
Synonym Pentanoic-2,2,3,3-d4 acid, Pentanoic acid-2,2,3,3-d4, Valeric acid-d4, Valeric-α,α,β,β-d4 acid, Pentanoic-2,2,3,3-D4 acid
InChI formel InChI=1 S/C5H10O2/c1-2-3-4-5(6)7/h2-4H2,1H3,(H,6,7)/i3D2,4D2
SMIL [2 H]C([2 H])(CC)C([2 H])([2 H])C(=O)O
IUPAC navn 2,2,3,3-tetradeuteriopentanoic acid
Molekylvægt (g/mol) 106.156
Formel vægt 106.093 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity